Order Entry
Canada
ContactUsLinkComponent
B-KER2® Laboratory Jar, Phipps & Bird
B-KER2® Laboratory Jar, Phipps & Bird
Catalog # 58968-220
Supplier:  Phipps & Bird
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
B-KER2® Laboratory Jar, Phipps & Bird
Catalog # 58968-220
Supplier:  Phipps & Bird
Supplier Number:  7790-630
Minority or Woman-Owned Business Enterprise
Small Business Enterprise

Specifications

  • Capacity:
    2000 ml (67628 oz.)
  • Description:
    Square, Laboratory Floc Jar
  • Cat. no.:
    58968-220
  • Supplier No.:
    7790-630

Specifications

About this item

The B-KER2® Jar is molded of strong, durable 1/4” thick acrylic

Its square shape provides thorough mixing action and in many cases accurately duplicates actual plant conditions. The jar comes complete with a sampling valve. It is compatible with two, four and six paddle jartesters, as well as most other stirrers. The jars are used to draw the jartest samples with a minimum of disturbance to the settling floc, it has a sampling port conveniently located at the 10 cm settling-distance level. There is no need to raise JarTester paddles.