ContactUsLinkComponent
Thermogreen® Injector Septa, PerkinElmer
Thermogreen® Injector Septa, PerkinElmer
Catalog # CA97013-078
Supplier:  PerkinElmer
undefined
Thermogreen® Injector Septa, PerkinElmer
Catalog # CA97013-078
Supplier:  PerkinElmer
Supplier Number:  N6621028
Specifications
  • Ø:
    11 mm
  • Description:
    Injector septa
  • Cat. no.:
    CA97013-078
  • Supplier No.:
    N6621028
Specifications
About this item
Thermogreen® Injector Septa are extremely low bleed over a wide range of inlet temperatures from 100 to 350 °C.
  • Functional over a wide range of temperatures
Easier needle penetration and high puncture tolerance make this septum ideal for autosamplers. Septa are already conditioned and ready to use.