Order Entry
Canada
ContactUsLinkComponent
Merit™ Probe and Excavator, Physician Grade, Sklar
Merit™ Probe and Excavator, Physician Grade, Sklar
Catalog # 10185-158
Supplier:  Sklar
undefined
Merit™ Probe and Excavator, Physician Grade, Sklar
Catalog # 10185-158
Supplier:  Sklar
Supplier Number:  98-508

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Length:
    152 mm (6")
  • Material:
    Stainless steel
  • Cat. no.:
    10185-158
  • Supplier No.:
    98-508

Specifications

About this item

Merit™ Probe and Excavator is industry standard.