Order Entry
Canada
ContactUsLinkComponent
Probe and Packer, OR Grade, Sklar
Probe and Packer, OR Grade, Sklar
Catalog # 10184-706
Supplier:  Sklar
undefined
Probe and Packer, OR Grade, Sklar
Catalog # 10184-706
Supplier:  Sklar
Supplier Number:  97-0539
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Straight and Sharp Probe and Packer
  • Length:
    152 mm (6")
  • Material:
    Stainless steel
  • Cat. no.:
    10184-706
  • Supplier No.:
    97-0539

Specifications

About this item

Probe and Packer is industry standard.