Order Entry
Canada
ContactUsLinkComponent
 
Northbent Suture Scissors, OR Grade, Sklar
  89025-338
 :  Sklar
undefined
Northbent Suture Scissors, OR Grade, Sklar
  89025-338
 :  Sklar
 :  22-2947
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

 

  • Description:
    Northbent Suture Scissors, Premium OR-Grade
  • Length:
    12 cm (4.75")
  • Material:
    Stainless steel
  • Environmentally Preferable:
  • Curvature:
    Curved
  • Cat. no.:
    89025-338
  • Sterility:
    Non sterile
  • Supplier No.:
    22-2947
  • Blade Type:
    Blunt/Blunt, Smooth
  • Handle:
    Finger Rings

 

 

Northbent Suture Scissors are primarily used for suture removal and not be used for cutting tissue.

These scissors have a small hook shaped tip on one blade, that slides under sutures to slightly lift them slightly before cutting for removal

Censitrac Ready products are industry standard.

This hook also holds the suture so it does not slip off the blade of the scissors prior to cutting. Northbent suture scissors have a light pattern, useful for suture removal.

 : Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
 : Customers must have a Censitrac Site ID to purchase Censitrac instruments.