QC Slide, Hardy Diagnostic
Catalog # CA10127-478
Supplier: HARDY DIAGNOSTICS CA
Specifications
- Description:Quartz slides Gram
- Cat. no.:CA10127-478
Specifications
About this item
A convenient ready-to-stain control slide that will test both your stain solutions and your staining techniques.
Positive and negative stain control organisms are affixed to the circles in each Q-Slide™. Up to four patient specimens (in the squares) may be processed at the same time as the QC organisms.