Poly HRP Dilution Buffer, Thermo Scientific
Catalog # CAPIN501
Supplier: Thermo Scientific
CAS Number:
Specifications
- Size:1 L
- Cat. no.:CAPIN501
- Supplier No.:N501
Specifications
About this item
Thermo Scientific Pierce Poly-HRP Dilution Buffer is optimized to ensure optimal performance with Poly-HRP Streptavidin.
Features of Poly-HRP Dilution Buffer:
Formulation: 1% biotin-free casein in a PBS buffer solution.
Application: Dilution of Poly-HRP Streptavidin (Product # N200): Gently invert bottle and swirl to mix before use. Do not vortex. Use at full strength. Refer to Poly-HRP Streptavidin Certificate of Analysis for recommended dilution ranges. Dilute Poly-HRP Streptavidin immediately before use.
Formulation: 1% biotin-free casein in a PBS buffer solution.
For Research Use Only. Not for use in diagnostic procedures.
Ordering information: Shipped on cold packs. Store at 2-80˚C immediately upon receipt.