Use the Thermo Scientific® Rat IFN gamma ELISA Kit to assay IFN-γ (rat interferon gamma) levels in rat serum, plasma (with EDTA, heparin, sodium citrate), and cell culture supernatant samples. This is an alternate product for Cat. No. KRC4021.
This kit uses pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of IFNγ levels in samples. The Thermo Scientific® Rat IFN gamma ELISA Kit also includes calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution, and stop solution. This complete kit contains one pre-coated 96-well strip plate and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.
Highlights:
Target: natural and recombinant rat interferon gamma (IFNγ)
Assay range: 8 to 500 pg/mL
- Primary antibody reactivity:Rat
- Target protein:IFN gamma