To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Assay Type:Competitive
- Target protein:Leukotriene B4
- Size:96 assays
- Cat. no.:CAPIEHLTB4
- Supplier No.:EHLTB4
Assay LTB4 (leukotriene B4) eicosanoid levels in biological samples, such as tissue culture media, plasma, saliva and urine, with our Thermo Scientific Leukotriene B4 Competitive ELISA Kit. This complete kit contains pre-coated antibody-plate, detection antibodies, diluent, wash buffer, standard, substrate, and stop solution.
Features of the Leukotriene B4 ELISA Kit:
Target: leukotriene B4 (LTB4, LT-B4)
Format: colorimetric ELISA (p-NPP substrate) in clear 96-well plate
Measurement: absorbance at 405nm minus absorbance at 570nm
Assay Range: 47 to 12,000 pg/mL
Sensitivity: < 19.4 pg/mL
Precision: dependent upon sample type and level (see instructions)
Sample Types: cell culture supernatant, serum, plasma (EDTA), saliva, urine
Total Assay Time: 3 hours
Sample Size: 100 µL
Specificity: no significant cross-reaction with PGE2,PGF2α, 20-OH-LTB4, 20-COOH-LTB4, LTC4, LTD4, LTE4, 5(S)-HETE, 12(S)-HETE, 15(S)-HETE; less than 1% cross-reaction with 12-epi-LTB4; approx. 5% cross-reaction with 6-trans-LTB4 and 6-trans-12-epi-LTB4.
This LTB4 competitive ELISA kit is an immunoassay for the quantitative determination of leukotriene B4 (LTB4) in biological fluids. The assay is based on the competition between LTB4 in the standard or sample and alkaline phosphatase conjugated LTB4 (LTB4-AP) for a limited amount of LTB4 monoclonal antibody bound to an anti-Rabbit IgG precoated 96-well plate. As the concentration of LTB4 in the sample increases, the amount of LTB4-AP captured by the coating antibody decreases. Thus, there is an inverse relationship between optical density (OD) and the amount of analyte in the sample.
Target: leukotriene B4 (LTB4, LT-B4)
Anti-Rabbit IgG Precoated 96-well Plate, 1 each
Reagent Diluent, 30 mL
LTB4 Antibody, 6 mL
LTB4-AP Conjugate, 6 mL
LTB4 Standard, 0.5 mL
10X Wash Buffer, 30 mL
Substrate Solution, 20 mL
Stop Solution, 6 mL
Plate Sealer, 1 each
LTB4 Assay Layout Sheet, 1 each