Pierce™ HisPur™ cobalt chromatography cartridges are convenient, ready-to-use pre-packed devices for efficient purification of polyhistidine-tagged proteins from bacterial, mammalian and baculovirus-infected cells.
- Binds at least 10 mg of purified 6 x His-tagged protein per ml of resin
- Cobalt-chelate coordination core binds fewer host protein contaminants, resulting in lower background than nickel resins
- Low metal leaching means no metal contamination in eluted histidine-tagged protein sample
HisPur™ cobalt chromatography cartridges are compatible with the major automated liquid-chromatography systems or manual syringe processing. The cartridges attach directly to ÄKTA™ or FPLC Systems without additional connectors. The included accessory pack readily adapts cartridges for use with Luer-Lok® syringe fittings or ¹/₁₆" tubing. These cartridges enable fast, easy and reproducible chromatographic separations and can be regenerated for multiple uses.
His-tagged proteins are purified from total soluble protein extract using a cobalt-charged tetradentate chelator immobilized onto 6% crosslinked agarose.
- Catalog No:
- CAPI90093
- CAPI90094
- Storage:
- Store between 4˚C and 8˚C.
- Store between 4˚C and 8˚C.