ContactUsLinkComponent
Pierce™ Immunoprecipitation, Crosslink Magnetic IP/Co-IP Kit, Thermo Scientific
Pierce™ Immunoprecipitation, Crosslink Magnetic IP/Co-IP Kit, Thermo Scientific
Catalog # CAPI88805
Supplier:  Thermo Scientific
CAS Number:  
Pierce™ Immunoprecipitation, Crosslink Magnetic IP/Co-IP Kit, Thermo Scientific
Catalog # CAPI88805
Supplier:  Thermo Scientific
Supplier Number:  88805
CAS Number:  
Specifications
  • Description:
    Crosslink Magnetic IP/Co-IP kit, Sufficient for 40 IP Reactions using 25 μL of Beads
  • Size:
    40 reactions
  • Cat. no.:
    CAPI88805
  • Supplier No.:
    88805
Specifications
About this item
The Thermo Scientific Pierce Crosslink Magnetic IP/Co-IP Kit uses crosslinking chemistry to covalently immobilize IP antibodies onto premium-quality Protein A/G Magnetic Beads for effective immunoprecipitation and co-immunoprecipitation.
  • Antibody is Irreversibly Attached to the Magnetic Beads Resulting in Negligible Co-Elution of Intact Antibody or Heavy and Light Chains with the Purified Antigen
  • Convenient – the Kit Contains Magnetic Beads and all Essential Buffers for Optimal Immunoprecipitation
  • Complete Crosslinking and Immunoprecipitation in Less than 3 Hours
  • Immunoprecipitate with Half the Recommended Volume of Magnetic Particles Compared to Other Magnetic Beads
  • Magnetic Beads Coupled with Protein A/G Recombinant Protein Ensures Compatibility with Most Primary Antibodies, Whether from Mouse or Rabbit
  • Use Only the Amount of Antibody Needed for a Single IP Experiment or Prepare a Larger Quantity of Antibody-Crosslinked Magnetic Beads for Multiple Experiments
Because this magnetic IP procedure involves covalent attachment of the specific antibody used for immunoprecipitation, target proteins or co-IP protein complexes can be eluted and analyzed without antibody fragments because the antibody remains affixed to the beads. The kit contains an optimized protocol and all buffers and reagents necessary to accomplish high-yield IP or co-IP with antibodies that bind to Protein A/G and using either manual or automated magnetic separation tools.
Delivery: The kit contains protein A/G magnetic beads, 1 mL; IP lysis/wash buffer, 2×50 mL; coupling buffer (20X), 25 mL; DSS crosslinker, 8×2 mg; lane marker sample buffer (5X), 5 mL; elution buffer, 25 mL; neutralisation buffer, 1 mLl; and lane marker sample buffer (5X), 5 mL Sufficient for 40 IP Reactions using 25 μL of Beads.
Caution: For Research Use Only. Not for use in diagnostic procedures. Store at 4˚C. Store DSS desiccated at 4˚C.