Pierce™ High Capacity Endotoxin Removal Spin Columns, Thermo Scientific
Supplier: Thermo Scientific
Certificates
About this item
Pierce™ high capacity endotoxin removal spin columns are ideal for the removal of endotoxins from protein or antibody samples prior to cell transfection or animal injection.
- Bind up to 2000000 EU/ml to eliminate grether than 99% of endotoxins
- Recover greater than 85% of protein sample
- Spin cup format enables endotoxin depletion within 1 hour
- Single use spin columns avoid cross contamination of samples
The Pierce™ high capacity endotoxin removal resin is a porous cellulose bead that has been surface-modified with covalently attached polylysine chains that have a high affinity for LPS but lacks the toxicity of other endotoxin-binding molecules.
Ordering information: Upon receipt store at 4 °C.
Caution: For research use only. Not for use in diagnostic procedures.
Related Information
Customers Who Bought This Also Bought