Dimethyl sulfoxide ≥99.5% for LC-MS, Pierce™
CAPI85190
:
- Size:50 ml
- Cat. no.:CAPI85190
- Supplier No.:85190
- Visual Appearance:Clear, Colorless liquid, Free of particulate matter
- Purity:≥ 99.5%
- Water Content:≤ 0.2%
:
:78.14 g/mol
:189 °C (1013 hPa)
:18.5 °C
:1.101 g/cm³ (20 °C)
:87 °C
:67-68-5
:(CH₃)₂SO
:MFCD00002089