Pierce™ UltraLink™ Monomeric Avidin Resin, Affinity Chromatography Resin, Thermo Scientific
CAPI53146
:
- Description:Monomeric Avidin UltraLink™ Resin
- Size:5 mL
- Storage conditions:Upon receipt store at 4˚C.
- Cat. no.:CAPI53146
- Supplier No.:53146
UltraLink™ Monomeric Avidin resin is ideal for affinity purification of biotinylated proteins, peptides and other molecules
- Non-denaturing purifies biotinylated products using mild elution conditions (2 mM free biotin)
- Reusable at least 10 times, with marginal loss in biotin binding capacity (approximately 2.5% decrease per regeneration)
- Retains biotin-binding specificity and low non specific binding of native avidin
- Binding capacity is >1.2 mg biotinylated BSA/mL resin
Immobilization of avidin monomers results in a purification resin with a much lower biotin-binding affinity than native tetrameric avidin, enabling recovery of biotinylated molecules using mild elution conditions.
: For Research Use Only. Not for use in diagnostic procedures.