ContactUsLinkComponent
Pierce™ 1-Step™ Ultra TMB-Blotting Solution, Thermo Scientific
Pierce™ 1-Step™ Ultra TMB-Blotting Solution, Thermo Scientific
Catalog # CAPI37574
Supplier:  Thermo Scientific
CAS Number:  
undefined
Pierce™ 1-Step™ Ultra TMB-Blotting Solution, Thermo Scientific
Catalog # CAPI37574
Supplier:  Thermo Scientific
Supplier Number:  37574
CAS Number:  
Specifications
  • Description:
    1-Step™ Ultra TMB-Blotting Solution
  • Size:
    250 mL
  • Cat. no.:
    CAPI37574
  • Supplier No.:
    37574
Specifications
About this item
Thermo Scientific 1-Step Ultra TMB-Blotting Solution is an enhanced single-component horseradish peroxidase (HRP) substrate for Western blotting and immunohistochemistry.
  • Protein bands visible in less than one minute
  • Protein bands stable after membrane drying
  • No special equipment needed for visualization, produces dark blue bands
  • Ready to use and sensitive
This precipitating, colorimetric Western blot substrate for HRP provides high sensitivity, increased signal-to-noise ratio and low background compared to many other chromogenic substrates. The blotting solution contains soluble TMB (3,3',5,5'-tetramethylbenzidine), which reacts very quickly with horseradish peroxidase enzyme to produce an insoluble dark blue precipitate that does not fade or flake. The substrate is compatible with both nitrocellulose and PVDF membranes.
Caution: For Research Use Only. Not for use in diagnostic procedures. Upon receipt store product at 4˚C.