Pierce™ DAB Substrate Kit, Thermo Scientific
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
The Thermo Scientific Pierce DAB Substrate Kit enables chromogenic detection of horseradish peroxidase (HRP) activity based the action of 3,3'-diaminobenzidine (DAB) in Western blot and tissue staining methods.
- Easy-to-use—dilute 10X stock solution to 1X in provided stable peroxide buffer
- Safer—preformulated DAB solution minimizes safety concerns with this chemical
- Complete—includes convenient solution of stable hydrogen peroxide buffer
- Store kit at 4˚C.
DAB (3,3'-diaminobenzidine tetrahydrochloride; MW = 214.1) reacts with HRP in the presence of peroxide to yield an insoluble brown-colored product at locations where peroxidase-conjugated antibodies are bound to samples. The brown precipitate is insoluble in alcohol and other organic solvents, making it an excellent substrate for immunohistochemical staining that requires the use of traditional counterstains and mounting media. The Pierce DAB Substrate Kit includes a 10X solution of DAB and Stable Peroxide Substrate Buffer. Crystalline DAB (powder) is also available.
Delivery information: Kit contains DAB Solution (10X), 25 ml; Stable Peroxide Substrate Buffer, 250 ml.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Documents
Specifications
Specifications
- Description:DAB Substrate Kit, sufficient for 25 uses at 10 mL each
- Size:275 mL
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



