ContactUsLinkComponent
Pierce™ 20X TAE Buffer, Thermo Scientific
Pierce™ 20X TAE Buffer, Thermo Scientific
Catalog # CAPI28354
Supplier:  Thermo Scientific
CAS Number:  
undefined
Pierce™ 20X TAE Buffer, Thermo Scientific
Catalog # CAPI28354
Supplier:  Thermo Scientific
Supplier Number:  28354
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    TAE buffer, 20X
  • Size:
    500 mL
  • Cat. no.:
    CAPI28354
  • Supplier No.:
    28354
Specifications
About this item
Thermo Scientific Pierce 20X TAE Buffer is a stock solution that is ideal for casting and preparing Tris-Acetate EDTA (pH 8.3) running buffer used for agarose gel electrophoresis of nucleic acids.
  • When diluted to 1X with water, yields 0.04M Tris, 0.04M acetate, 1 mM EDTA, pH 8.2 to 8.4
  • Easy to use
  • Saves time and space
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area.
Caution: For Research Use Only. Not for use in diagnostic procedures.