Order Entry
Canada
ContactUsLinkComponent
PageRuler™ Prestained NIR Protein Ladder
PageRuler™ Prestained NIR Protein Ladder
Catalog # CAPI26635
Supplier:  Thermo Scientific
CAS Number:  
PageRuler™ Prestained NIR Protein Ladder
Catalog # CAPI26635
Supplier:  Thermo Scientific
Supplier Number:  26635
CAS Number:  

Specifications

  • Size:
    2 x 250 µl
  • Storage conditions:
    Upon receipt store at -20˚C
  • Protein/peptide name:
    Prestained NIR Protein Ladder
  • Shipping temperature:
    Product is shipped with an ice pack.
  • Cat. no.:
    CAPI26635
  • Supplier No.:
    26635

Specifications

About this item

Thermo Scientific PageRuler Prestained NIR Protein Ladder is a mixture of 10 proteins (11 to 250KDa) that are blue-stained and fluor-labeled for near-IR fluorescent visualization and protein sizing following electrophoresis.

Features of PageRuler Prestained NIR Protein Ladder:

Size range—10 proteins spanning 11 to 250kDa
Fluorescent—detect using 670nm red laser or 700nm channels with fluorescence imagers
Ready-to-use—supplied in a loading buffer for direct loading on gels
Sharp bands—excellent sizing for near-infrared visualization of protein gels and blots
Reference band—55kDa band has greater intensity for easy orientation
Quality tested—each lot evaluated by SDS-PAGE and Western blotting

The protein MW markers in this ladder resolve into sharp bands when analyzed by SDS-PAGE and are labeled with a fluorescent dye for visualization with instruments equipped for detection of near-infrared (NIR) fluorescence. The bands are also directly visible because the proteins are prestained blue. The 55kDa band is of greater intensity and serves as a reference band. The protein ladder is conveniently packaged and ready to use: no heating, dilution or addition of a reducing agent is required before use.

Includes:
Proteins in 62.5 mM Tris-H3PO4 (pH 7.5 at 25˚C), 1 mM EDTA, 2% SDS, 10 mM DTT, 1 mM NaN3 and 33% glycerol

Ordering information: For Research Use Only. Not for use in diagnostic procedures.