- Size:2 x 250 µl
- Storage conditions:Upon receipt store at -20˚C
- Protein/peptide name:Multicolor High Range Protein Ladder
- Shipping temperature:Product is shipped with an ice pack.
- Cat. no.:CAPI26625
- Supplier No.:26625
Thermo Scientific Spectra Multicolor High Range Protein Ladder is a mixture of eight (8) blue-, green-and orange-stained proteins (40 to 300kDa) for use as size standards for high-MW proteins in gel electrophoresis and Western blotting.
Features of Spectra Multicolor High Range Protein Ladder:
Size range—eight proteins spanning 40 to 300kDa
Multicolor—three different colors for unambiguous band-size assignment
Ready-to-use—supplied in a loading buffer for direct loading on gels; no need to boil
Sharp bands—color-coded bands of similar intensity for easy visualization
Quality tested—each lot evaluated by SDS-PAGE and Western blotting
Membrane-compatible—colored bands transfer to membranes for Western blotting
Includes:
Dye-stained proteins in 62.5 mM Tris-H3PO4 (pH 7.5 at 25˚C), 1 mM EDTA, 2% SDS, 10 mM DTT, 1 mM NaN3 and 33% glycerol.
Applications:
Monitoring of protein migration during SDS-PAGE
Verifying Western transfer efficiency
Approximate sizing of proteins on SDS-polyacrylamide gels and Western blots
Locating a protein of interest for excision from an unstained preparative gel