Order Entry
Canada
ContactUsLinkComponent
PageRuler™ Unstained Protein Ladder
PageRuler™ Unstained Protein Ladder
Catalog # CAPI26614
Supplier:  Thermo Scientific
CAS Number:  
PageRuler™ Unstained Protein Ladder
Catalog # CAPI26614
Supplier:  Thermo Scientific
Supplier Number:  26614
CAS Number:  

Specifications

  • Size:
    2 x 250 µl
  • Storage conditions:
    Upon receipt store at -20˚C. Product with an ice pack.
  • Protein/peptide name:
    Unstained Protein Ladder
  • Cat. no.:
    CAPI26614
  • Supplier No.:
    26614

Specifications

About this item

Thermo Scientific PageRuler Unstained Protein Ladder is a mixture of 14 recombinant, highly purified, unstained proteins (10kDa to 200kDa) for use as size standards in protein electrophoresis (SDS-PAGE) and Western blotting.

Features of PageRuler Unstained Protein Ladder:

Size range—14 proteins spanning 10 to 200kDa
Ready-to-use—supplied in a loading buffer for direct loading on gels
Sharp bands—excellent accuracy for detection in with coomassie or silver stains
Reference band—50kDa band is has greater intensity for easy orientation
Tagged—each protein in the ladder contains an integral Strep-tag™ II Sequence
Quality tested—each lot evaluated by SDS-PAGE and Western blotting

Each protein in the ladder contains an integral Strep-tag™ II Sequence which can be detected directly on Western blots using a Strep-Tactin™ Conjugates or an antibody against Strep-tag™ II Sequence (see Refs 2-3). The Ladder produces sharp bands on SDS-polyacrylamide gel following staining of the gel with Thermo Scientific Pierce Gel Stains or other coomassie dye or silver stains. The 50kDa protein band is of greater intensity and serves as a reference band.

Includes:
Proteins (0.02 to 0.05 mg/mL of each) in 62.5 mM Tris-H3PO4 (pH 7.5 at 25˚C), 1 mM EDTA, 2% SDS, 10 mM DTT, 1 mM NaN3, 0.01% bromophenol blue and 33% glycerol

Ordering information: For Research Use Only. Not for use in diagnostic procedures.