Pierce® Pegylation Reagents, Methyl-PEG-Thiol Compound, Thermo Scientific
Catalog # CAPI26132
Supplier: Thermo Scientific
CAS Number:
Specifications
- Description:MT(PEG)4
- Size:100 mg
- Storage conditions:Upon receipt store at -20˚C protected from moisture.
- Cat. no.:CAPI26132
- Spacer Arm:16 Å
- Supplier No.:26132
Specifications
About this item
Methyl-PEG4-thiol is a methyl- and sulfhydryl-terminated compound that contains a 4-unit polyethylene glycol (PEG) spacer and is used to modify surfaces such as quantum dots, monolayers and magnetic particles.
- PEG groups are flexible, non-immunogenic and hydrophilic
- Terminal monodentate thiol reacts spontaneously with gold surfaces through dative binding
- Terminal methyl group coats quantum dot and other surfaces to reduce nonspecific binding in assays
The use of MT(PEG)4 with CT(PEG)12 in surface modification can form a hydrophilic 'lawn' of methyl ether-terminated PEGs with periodic exposed carboxylic acid-containing PEGs. The exposed carboxylic acid groups can be coupled to affinity ligands using the carbodiimide coupling reaction with EDC and sulfo-NHS.
Caution: For Research Use Only. Not for use in diagnostic procedures.