ContactUsLinkComponent
Pierce™ Streptavidin, Maleimide-Activated, Thermo Scientific
  CAPI21102
 :  
Pierce™ Streptavidin, Maleimide-Activated, Thermo Scientific
  CAPI21102
 :  21102
 :  
  • Description:
    Streptavidin, Maleimide-Activated
  • Size:
    1 mg
  • Storage conditions:
    Upon receipt store at room temperature or 4˚C
  • Cat. no.:
    CAPI21102
  • Supplier No.:
    21102
Thermo Scientific Pierce Maleimide-Activated Streptavidin conjugate include streptavidin in a purified form, activated for crosslinking to sulfhydryl groups containing molecules.
  • Provided as a lyophilized powder containing sodium citrate
  • Facilitates conjugation of Streptavidin to proteins
Maleimide-activated Streptavidin is designed to facilitate the conjugation of the Streptavidin to proteins, peptides and ligands that contain sulfhydryl (-SH) groups. The reactive maleimide group that has been added to the Streptavidin allows conjugation reactions to be carried out in the pH range 6.5-7.5, creating a stable thio-ether link.
 : For Research Use Only. Not for use in diagnostic procedures.