Order Entry
Canada
ContactUsLinkComponent
Pierce™ D-Galactose Agarose, Thermo Scientific
Pierce™ D-Galactose Agarose, Thermo Scientific
Catalog # CAPI20372
Supplier:  Thermo Scientific
CAS Number:  
undefined
Pierce™ D-Galactose Agarose, Thermo Scientific
Catalog # CAPI20372
Supplier:  Thermo Scientific
Supplier Number:  20372
CAS Number:  

Specifications

  • Description:
    D-Galactose Agarose
  • Size:
    5 mL
  • Binding capacity:
    >20 mg Jacalin per mL of Settled Resin
  • Cat. no.:
    CAPI20372
  • Supplier No.:
    20372

Specifications

About this item

Thermo Scientific Pierce D-Galactose Agarose enables purification of lectins, galactosidases and other galactose-binding molecules.

  • Ligand: thio-alpha-D-galactose
  • Synonyms: thio-alpha-gal, thio-gal
  • Support: crosslinked 6% beaded agarose
  • Format: 50% resin slurry in 0.02% sodium azide
  • Storage: 4˚C

The Immobilized D-galactose is a beaded agarose affinity resin that enables purification of agglutinins, lectins, toxins, and other galactose-binding proteins, N-acetylgalactosamine-binding proteins or carbohydrate-binding proteins. This purification method is milder and/or more specific than other methods, allowing protein recovery in a manner that preserves activity for downstream applications.

Caution: For Research Use Only. Not for use in diagnostic procedures.