Ethidium bromide solution 0,625 mg/mL, Pierce™
Product Details & Documents
Thermo Scientific Pierce Ethidium Bromide is available as a dilute and concentrated solution to accommodate most molecular biology applications for fluorescent DNA visualization.
Ethidium Bromide is an intercalating dye used in the visualization of DNA and RNA in electrophoresis gels and cesium chloride density gradients.
Caution: For Research Use Only. Not for use in diagnostic procedures.
- CAS Number:1239-45-8
- Formula:C₂₁H₂₀BrN₃
- MDL Number:MFCD00011724
- Molecular Weight:394.31 g/mol
Documents
Specifications
Specifications
- Size:5 mL
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



