ContactUsLinkComponent
Pierce™ Luciferase Cell Lysis Buffer (2X), Thermo Scientific
  CAPI16189
 :  
undefined
Pierce™ Luciferase Cell Lysis Buffer (2X), Thermo Scientific
  CAPI16189
 :  16189
 :  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Luciferase cell lysis buffer, 2X
  • Size:
    250 mL
  • Storage conditions:
    Store at room temperature
  • Cat. no.:
    CAPI16189
  • Supplier No.:
    16189
The Thermo Scientific Pierce Luciferase Cell Lysis Buffer effectively lyses adherent cells or pelleted suspension cells for analysis of promoter expression in luciferase assays.
  • Compatible with all Pierce Luciferase Reporter Assays
  • Enables rapid and efficient lysis
  • Tested compatibility with multiple Mammalian cell lines
  • Compatible with protein assays (e.g., Coomassie Plus and BCA)
Pierce Luciferase Cell Lysis Buffer has been tested and validated to produce luciferase assay-compatible lysates from a variety of cell types. Essentially complete lysis of adherent cells has been observed with nearly all of the cell types tested to date, including COS-7, NIH3T3, Hepa 1-6, 293, CHO, MDA, MB 231, Jurkat, and FM2 cells.
 : For Research Use Only. Not for use in diagnostic procedures.