Order Entry
Canada
Orders LinkContactUsLinkComponent
Rabbit IgG Isotype Control

Rabbit IgG Isotype Control

1.0 mg
Catalog # 10014-036
Supplier:   Immunoreagents
Supplier #:  RB-003-V
Rabbit IgG Isotype Control
Catalog # 10014-036
Supplier:  Immunoreagents
Rabbit IgG Isotype Control

1.0 mg

Price
$70.80
Log in for availability
Each (1mg)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

ImmunoReagents is a manufacturer of quality research and production reagents for the Life Science Industries. Product lines include a wide range of antibodies used in ELISA, immunohistochemistry (IHC), western blotting, flow cytometry (FACS), immunofluorescence and microscopy.  Antibodies produced cover a broad range of Immunoglobulins from various species cross absorbed to provide high specificity.  

  • Host: Rabbit
  • Conjugation: Unconjugated
  • Clonality: Polyclonal
  • Isotype: IgG
  • Reactivity: Rabbit IgG
  • Concentration: 1 mg/mL (E 1% at 280 nm = 14.0)
  • Immunogen: Purified Rabbit IgG, whole molecule
  • Purification Method: Affinity Purified
  • Form: Liquid, 0.2 µm filtered
  • Storage Buffer: 2 mM Sodium Phosphate, 0.03 M Sodium Chloride, pH 7.8, 10 mg/mL Sucrose
  • Storage Temperature: 2 to 8°C
  • Shipping Temperature: 2 to 8°C
  • Format: Purified IgG

For use as a standard or control in most immunoassay formats.

Specifications
  • Size:
    1.0 mg
  • Clonality:
    Polyclonal
  • Purification:
    Affinity Purified
  • Antibody type:
    Control
  • Form:
    Liquid, 0.2 µm filtered
  • Isotype:
    IgG
  • Storage temperature:
    2 to 8°C
  • Host:
    Rabbit
  • Concentration:
    1 mg/mL (E 1% at 280 nm = 14.0)
  • Shipping temperature:
    2 to 8°C
  • Cat. No.:
    10014-036
  • Reactivity:
    Rabbit
  • Format:
    Purified IgG
  • Immunogen:
    Purified Rabbit IgG, whole molecule
  • Conjugation:
    Unconjugated
  • Storage buffer:
    2 mM Sodium Phosphate, 0.03 M Sodium Chloride, pH 7.8, 10 mg/mL Sucrose