NWCP Nitrile Co-Polymer DI Water Washed Class 10 Gloves, CT International
Supplier: CT International
Certificates
About this item
NWCP nitrile co-polymer DI water washed class 10 gloves are manufactured for outstanding strength, stretch and fit.
- No latex proteins
- Anti-static
- Double bagged
- Tapered, beaded cuff
- Leak tested
They offer textured finger tips for critical handling and a super soft feel for comfort.
Certifications: NWCP series class 10 nitrile gloves are ISO 4 cleanroom compatible or higher controlled environments.
Specifications
- Catalog No:
- 56613-720
- 56613-724
- 56613-728
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Cleanroom class:
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- ASTM No.:
- D3578-05
- D3578-05
- D3578-05