Formaldehyde Solution Spill Kit
Formaldehyde Solution Spill Kit
470219-190
: IS5035
Be prepared for spills in your lab.
- Enough Material to Clean One Liter of a 10% Formalin Solution
- Instructions and SDS
- Vinyl Exam Gloves
This kit includes a dust pan and brush to properly dispose of spills.
It also includes vinyl exam gloves, so you are protected while cleaning.
: This kit includes an instruction sheet/SDS, vinyl exam gloves, polybags, twist ties, blank shipping tags, dust pan and brush, vermiculite, and an absorbent formaldehyde neutralizing agent (sodium bisulfite).
- Description:Formaldehyde Solution Spill Kit