Orders LinkContactUsLinkComponent
Two-Piece Tissue Cassettes
Two-Piece Tissue Cassettes
Two-piece tissue cassettes, White
Catalog # 10022-344
Supplier:   VWR International
Two-Piece Tissue Cassettes
Catalog # 10022-344
Supplier:  VWR International
Two-Piece Tissue Cassettes
Two-Piece Tissue Cassettes
Two-piece tissue cassettes, White
Price
$224.76
New Customer Promotional Price
Log in for availability
Case of 500
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Two-Piece Tissue Cassettes are molded from a special, high-density polymer that is resistant to the chemical actions of solvents.
  • Three frosted writing areas
  • Two-piece design
  • Lid relocks without risk of specimen loss
  • Chemical-resistant
Lids can be snapped on and off as often as necessary and relock without danger of specimen loss.
Cassettes have a two-piece design and feature three frosted labeling areas.
Packaging: 500 per case.
Specifications
  • Color:
    White
  • Description:
    Two-piece tissue cassettes
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)