Halogen Bulbs, Ushio, Bulbtronics©
Supplier: Bulbtronics
Jump to:
About this item
Various halogen bulbs.
15 Options Available Below
Product Details & Documents
Various halogen bulbs.
Product Family Options
Product Information
- FilamentBulb lifeLengthØModelBaseVoltagePowerOEM numberAvailabilityPrice
- 47739-622Bulb, Ushio, 19.56 mm light center lengthBulb, Ushio, 19.56 mm light center lengthC6/single coil transverse100 h30 mm8.89 mmT2 3/4G4/2-pin6 V20 W1000532Log in for availabilityEach$31.55Supplier Number: 0001507
- 47740-234Bulb, Ushio, 19.56 mm light center lengthBulb, Ushio, 19.56 mm light center lengthC6/single coil transverse100 h30 mm8.89 mmT2 3/4G4/2-pin6 V10 W1000531Log in for availabilityEach$241.37Supplier Number: 0001813
- 47740-892Bulb, Ushio, 19.3 mm light center lengthBulb, Ushio, 19.3 mm light center lengthC6/single coil transverse50 h30 mm8.89 mmT2 3/4G4/2-pin6 V30 W1000867Log in for availabilityEach$62.54Supplier Number: 0002142
- 89041-226Bulb, Ushio, 19.56 mm light center lengthBulb, Ushio, 19.56 mm light center lengthC6/single coil transverse2000 h30 mm8.89 mmT2 3/4G4/2-pin6 V20 W1000866Discontinued-Supplier Number: -
- 47740-886Bulb, Ushio, 19.56 mm light center lengthBulb, Ushio, 19.56 mm light center lengthC6/single coil transverse300 h31 mm7.87 mmT2 1/2G4/2-pin6 V10 W1000857Log in for availabilityEach$239.55Supplier Number: 0002139
- 47740-974Bulb, Ushio, stippled dichroic reflectorBulb, Ushio, stippled dichroic reflectorC6/single coil transverse50 h33.27 mm34.93 mmMR11GZ4/2-pin6 V20 W1000934Log in for availabilityEach$172.92Supplier Number: 0002183
- 47740-484Bulb, Ushio, Faceted dichroic reflectorBulb, Ushio, Faceted dichroic reflectorC6/single coil transverse2000 h35 mm34.93 mmMR11GZ4/2-pin12 V20 W1000621Discontinued-Supplier Number: 0001938
- 47739-214Bulb, Ushio, Specular dichroic reflectorBulb, Ushio, Specular dichroic reflectorC8/single coil axial50 h41.91 mm50.8 mmMR16GZ6.35/2-pin15 V150 W1000272Log in for availabilityEach$100.94Supplier Number: 0001303
- 47738-848Bulb, Ushio, 30 mm light center lengthBulb, Ushio, 30 mm light center lengthCBAR6/C6F/single coil flat/oval transverse50 h44 mm11.43 mmT3 1/2G6.35/2-pin12 V50 W1000071Discontinued-Supplier Number: -
- 47739-724Bulb, Ushio, 30 mm light center lengthBulb, Ushio, 30 mm light center lengthCBAR6/C6F/single coil flat/oval transverse2000 h44 mm11.43 mmT3 1/2GY6.35/2-pin12 V100 W1000381Discontinued-Supplier Number: -
- 47740-088Bulb, Ushio, 30 mm light center lengthBulb, Ushio, 30 mm light center lengthCBAR6/C6F/single coil flat/oval transverse50 h44 mm10.92 mmT3 1/2GY6.35/2-pin12 V100 W1000490Each$29.94Pack of 10$299.40Supplier Number: 0001740
- 47738-988Bulb, Ushio, Faceted dichroic reflectorBulb, Ushio, Faceted dichroic reflectorCC6/coiled coil transverse500 h44.45 mm50.8 mmMR16GX5.3/2-pin20 V150 W1000173Log in for availabilityEach$114.37Supplier Number: 0001190
- 47739-350Bulb, Ushio, Specular dichroic reflectorBulb, Ushio, Specular dichroic reflectorCC6/coiled coil transverse40 h44.45 mm50.8 mmMR16GX5.3/2-pin21 V150 W1000297Log in for availabilityEach$100.24Supplier Number: 0001371
- 47739-394Bulb, Ushio, Specular dichroic reflectorBulb, Ushio, Specular dichroic reflectorCC6/coiled coil transverse40 h44.45 mm50.8 mmMR16GX5.3/2-pin21 V150 W1000302Log in for availabilityEach$109.52Supplier Number: 0001393
- 47739-398Bulb, Ushio, Specular dichroic reflectorBulb, Ushio, Specular dichroic reflectorCC6/coiled coil transverse25 h44.45 mm50.8 mmMR16GX5.3/2-pin19 V80 W1000303Discontinued-Supplier Number: -
- 47739-414Bulb, Ushio, Specular dichroic reflectorBulb, Ushio, Specular dichroic reflectorCC6/coiled coil transverse200 h44.45 mm50.8 mmMR16GX5.3/2-pin21 V150 W1000306Log in for availabilityEach$66.77Supplier Number: 0001403
- 47739-430Bulb, Ushio, Specular dichroic reflectorBulb, Ushio, Specular dichroic reflectorCC6/coiled coil transverse120 h44.45 mm50.8 mmMR16GX5.3/2-pin17.7 V120 W1000311Discontinued-Supplier Number: -
- 47739-436Bulb, Ushio, Specular dichroic reflectorBulb, Ushio, Specular dichroic reflectorCC6/coiled coil transverse200 h44.45 mm50.8 mmMR16GX5.3/2-pin10.8 V30 W1000315Log in for availabilityEach$132.11Supplier Number: 0001414
- 47739-456Bulb, Ushio, Specular dichroic reflectorBulb, Ushio, Specular dichroic reflectorCC6/coiled coil transverse50 h44.45 mm50.8 mmMR16GX5.3/2-pin24 V250 W1000318Discontinued-Supplier Number: -
- 47739-814Bulb, Ushio, stippled dichroic reflectorBulb, Ushio, stippled dichroic reflectorCC8/coiled coil axial35 h44.45 mm41.28 mmMR13GX5.3/2-pin82 V300 W1000414Discontinued-Supplier Number: -
- 47755-862Bulb, Ushio, 44.45 mm light center length, dichroic reflectorBulb, Ushio, 44.45 mm light center length, dichroic reflectorCC6/coiled coil transverse200 h50.8 mm50.04 mmMR16GX5.3/2-pin21 V150 W1001628Discontinued-Supplier Number: 0016423
- 47739-922Bulb, Ushio, 32 mm light center lengthBulb, Ushio, 32 mm light center lengthCC8/coiled coil axial75 h56.9 mm11.43 mmT3 1/2G5.3/2-pin82 V360 W1000442Each$60.73Pack of 5$303.65Supplier Number: 0001657
- 47739-160Bulb, Ushio, 36.58 mm light center lengthBulb, Ushio, 36.58 mm light center lengthCC6/coiled coil transverse75 h63 mm22.23 mmG7GY9.5/2-pin prefocus120 V600 W1000251Discontinued-Supplier Number: -
- 47739-962Bulb, UshioBulb, UshioCC6/coiled coil transverse150 h63.5 mm22.23 mmG7G5.3/2-pin120 V250 W1000453Log in for availabilityEach$185.54Supplier Number: 0001677
- 47739-714Bulb, Ushio, 35 mm light center lengthBulb, Ushio, 35 mm light center lengthCC8/coiled coil axial2000 h71.12 mm12.7 mmT4E11/MINI-Candelabra120 V150 W1000377Discontinued-Supplier Number: -
- 47740-062Bulb, UshioBulb, UshioCC8/coiled coil axial35 h80.26 mm14.99 mmT4 3/4R7S/RSC recessed single contact120 V650 W1000485Discontinued-Supplier Number: -
- 47739-358Bulb, UshioBulb, UshioC8/single coil axial400 h119.63 mm11.94 mmT3 3/4R7S/RSC recessed single contact120 V750 W1000299Log in for availabilityEach$70.42Supplier Number: 0001375
- 47740-568Bulb, UshioBulb, UshioC8/single coil axial100 h135.13 mm11.94 mmT3 3/4R7S/RSC recessed single contact120 V500 W1000652Discontinued-Supplier Number: 0001980