To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:1 mg
- Protein/peptide name:pp60v-src
- Purity:>98%
- Molecular weight:2482.7
- Molecular formula:C₁₁₁H₁₆₈N₃₀O₃₅
- Cat. no.:89143-664
- Supplier No.:165-004-M001
Specifications
About this item
Receptor tyrosine kinase inhibitor
H-Val-Ala-Pro-Ser-Asp-Ser-Ile-Gln-Ala-Glu-Glu-Trp-Tyr-Phe-Gly-Lys-Ile-Thr-Arg-Arg-Glu-OH
Inhibitor of Src and EGF receptor tyrosine kinases. The peptide sequence corresponds to residues 137-157 of the noncatalytic domain of the transforming protein of Rous sarcoma virus (pp60v-scr).