About this item
This ready-to-use Universal Antibody Dilution Buffer is designed for dilution of antibodies in all immunoassays, including immunofluorescence, IHC, ELISA, WB
- Use to dilute antibodies, primary or secondary as recommended in the protocol
- Diluted antibodies can be stored at 2 to 8 °C for several weeks, depending upon the stability of antibodies
This buffer does not contain any mammalian proteins, phosphate, sodium azide, or mercury preservatives and can be used for dilution of all antibodies, including peroxidase, and antibodies to phophoproteins. However, it is not suitable for dilution of antibodies to S100 proteins.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.