This ready-to-use Antibody Dilution Buffer is designed for the dilution of antibodies in all immunoassays, including immunofluorescence, IHC, ELISA, WB
- Ready to use buffer
- Use to dilute antibodies, primary or secondary as recommended in the protocol
- Diluted antibodies can be stored at 2 – 8°C for several weeks, depending upon the stability of antibodies
- Store at 2 to 8 °C
This buffer contains immunoglobulin free BSA and does not contain phosphate, sodium azide, or mercury preservatives. It can be used for the dilution of all antibodies, including peroxidase, and antibodies to phophoproteins. However, it is not suitable for dilution of antibodies to S100 proteins. This buffer contains green food color.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.