If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:Blue Silicone Rubber Kit
- Cat. no.:102092-300
- Supplier No.:24230-B
Specifications
About this item
Blue silicone rubber is extremely high tear, high tensile strength, two components and tin catalyzed RTV silicone rubber
- Low shrinkage and low viscosity
- Excellent chemical resistance
- Excellent shelf aging
- Color coded catalyst
- Long pot life insensitive to inhibition
- Variable cure rate
- Good dielectric properties
It is designed for casting polyurethane foam, polyester and epoxy parts.