ContactUsLinkComponent
Combined Hematoxylin and Eosin/Methenamine Silver Stain for the Histological Diagnosis of Fungi in Tissue, Electron Microscopy Sciences
Combined Hematoxylin and Eosin/Methenamine Silver Stain for the Histological Diagnosis of Fungi in Tissue, Electron Microscopy Sciences
Catalog # CA11028-246
CAS Number:  
undefined
Combined Hematoxylin and Eosin/Methenamine Silver Stain for the Histological Diagnosis of Fungi in Tissue, Electron Microscopy Sciences
Catalog # CA11028-246
Supplier Number:  26123-01
CAS Number:  
Specifications
  • Description:
    Chromic Acid 10%
  • Cat. no.:
    CA11028-246
  • Supplier No.:
    26123-01
Specifications
About this item
A Combined Hematoxylin and Eosin/Methenamine Silver Stain is used for the Histological Diagnosis of Fungi in Tissue Sections.
  • For bacterial, fungal stains
Stain results - Organism: Blue-Black Background: Rose.
Lamps, Laura W. et al A Combined Hematoxylin and Eosin/Methenamin Silver Stain for theHistological Diagnosis of Fungi In Tissue Sections., J. of Histotechnology 23:4 p.341 – 345, 2000.