Order Entry
Canada
Orders LinkContactUsLinkComponent
Sorensen's Phosphate Buffer, Electron Microscopy Sciences | Avantor

Sorensen's Phosphate Buffer, Electron Microscopy Sciences

Sorensen's Phosphate Buffer, 0.15 M, 500 ml, 2 x 0.5 mm Slot, —
Catalog # 100496-402
Minority or Woman-Owned Business Enterprise
Fournisseur #:  11682-05
Sorensen's Phosphate Buffer, Electron Microscopy Sciences
Catalog # 100496-402
Minority or Woman-Owned Business Enterprise
CAS Number:  
Sorensen's Phosphate Buffer, Electron Microscopy Sciences
Sorensen's Phosphate Buffer, Electron Microscopy Sciences

Sorensen's Phosphate Buffer, 0.15 M, 500 ml, 2 x 0.5 mm Slot, —

CAS Number:  
Price
$90.89
Log in pour la disponibilité
Each (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Sorensen's Phosphate Buffer is also know as phosphate mixed sodium salts. The osmolarity of a 0.1 M at a pH 7.2 is 226 mosmols; with the addition of 0.18 M of sucrose to the solution, the osmolarity raises to 425 mosmols.

  • pH can be specified when ordering
  • Maximum shelf life after receipt is 3 months

Ordering information: Certificates of analysis are available by lot or product number.

Spécifications

Spécifications

  • Description:
    Sorensen's Phosphate Buffer, 0.15 M
  • Size:
    2 x 0.5 mm Slot
  • pH:
  • Volume:
    500 ml
  • Cat. No.:
    100496-402
  • Size:
    500 ml

Spécifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.