Order Entry
Canada
ContactUsLinkComponent
 
Littauer Tapered Cutting Forceps, OR Grade, Sklar
Catalog # 10184-734
Supplier:  Sklar
undefined
Littauer Tapered Cutting Forceps, OR Grade, Sklar
Catalog # 10184-734
Supplier:  Sklar
Supplier Number:  97-1174

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Tapered Littauer Cutting Forceps
  • Length:
    12.7 cm (5")
  • Material:
    Stainless steel
  • Tip Configuration:
    Curved Back Blades, Smooth
  • Curvature:
    Straight
  • Cat. no.:
    10184-734
  • Supplier No.:
    97-1174

Specifications

About this item

Sklar's® Littauer Tapered Cutting Forceps are a double spring, plier instrument often used in podiatry as a nail nipper.The tapered blades form a narrow profile and sharp tips useful for accessing difficult to reach sections of a nail.

Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.