Order Entry
ContactUsLinkComponent
Anti-ZCCHC11 Goat Polyclonal Antibody
Anti-ZCCHC11 Goat Polyclonal Antibody
Catalog # CAPIPA5-17939
Supplier:  Thermo Scientific
Anti-ZCCHC11 Goat Polyclonal Antibody
Catalog # CAPIPA5-17939
Supplier:  Thermo Scientific
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Zinc Finger, CCHC Domain Containing 11
  • Antigen symbol:
    ZCCHC11
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Size:
    100 µg
  • Antigen synonyms:
    6030404K05Rik|Zinc finger CCHC domain containing 11|KIAA0191|PPAPD3|mKIAA0191|zinc finger CCHC domain-containing protein 11|9230115F04Rik|RP23-406B13.1|TUTase 4|terminal uridylyltransferase 4
  • Storage buffer:
    TBS, pH 7.3, with 0.5% BSA and 0.02% Sodium azide
  • Storage temperature:
    –20 °C, avoid freeze/thaw cycles
  • Cross adsorption:
    No
  • Tested applications:
    Immunohistochemistry
  • Cat. no.:
    CAPIPA5-17939
  • Supplier No.:
    PA5-17939

Specifications

About this item

This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology. ZCCHC11 is an RNA uridyltransferase that uses UTP to add uridines to the 3-prime end of substrate RNA molecules .

Recommended Dilutions: Immunohistochemistry: 2-6µg/ml

Type: Primary
Antigen: ZCCHC11 (zinc finger, CCHC domain containing 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human

Frequently Bought Together