Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin D
- Antigen symbol:IgD
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:500 µL
- Epitope:Heavy chain
- Cross adsorption:No
- Storage buffer:PBS, pH 7.4, with 0.2% BSA and 0.09% Sodium azide
- Storage temperature:4 °C
- Tested applications:Immunohistochemistry (Paraffin)
- Cat. no.:CAPIPA5-16400
- Supplier No.:PA5-16400
Specifications
About this item
This Antibody targets Human IgD (delta-Heavy Chain) in IHC (P) and WB applications and shows reactivity with Human samples. The immunogen is igD isolated from a pool of normal human sera. Thermo Scientific Pierce Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e. g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i. e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc. ) to generate highly specific reagents.
Recommended Dilutions: Western Blot: 1:100-1:1000, Immunohistochemistry (Paraffin): 1:500-1:1000
Type: Secondary
Antigen: Human IgD
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human