To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Ectonucleotide pyrophosphatase/phosphodiesterase 1
- Antigen symbol:ENPP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Cross adsorption:No
- Antigen synonyms:PCA1|phosphodiesterase I/nucleotide pyrophosphatase 1|M6S1|membrane component|surface marker 1|plasma-cell membrane glycoprotein 1|alkaline phosphodiesterase 1|ENPP1|HGNC:3356|PDNP1|Ly-41 antigen|PC-1|NPP1|NPPS|ectonucleotide pyrophosphatase/phosphodiesterase 1|OTTHUMP00000043194|chromosome 6
- Storage buffer:TBS, pH 7.3, with 0.5% BSA and 0.02% Sodium azide
- Storage temperature:–20 °C, avoid freeze/thaw cycles
- Tested applications:Immunohistochemistry
- Cat. no.:CAPIPA5-18429
- Supplier No.:PA5-18429
Specifications
About this item
This antibody is predicted to react with mouse and rat based on sequence homology. This gene is a member of the ecto-nucleotide pyrophosphatase/phosphodiesterase family. The encoded protein is a type II transmembrane glycoprotein comprising two identical disulfide-bonded subunits. This protein has broad specificity and cleaves a variety of substrates, including phosphodiester bonds of nucleotides and nucleotide sugars and pyrophosphate bonds of nucleotides and nucleotide sugars. This protein may function to hydrolyze nucleoside 5' triphosphates to their corresponding monophosphates and may also hydrolyze diadenosine polyphosphates. Mutations in this gene have been associated with 'idiopathic' infantile arterial calcification, ossification of the posterior longitudinal ligament of the spine , and insulin resistance.
Recommended Dilutions: Western Blot: 1-3 ug/ml, Immunohistochemistry: 1-2µg/ml
Type: Primary
Antigen: ENPP1 (ectonucleotide pyrophosphatase/phosphodiesterase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human