ContactUsLinkComponent
Merit™ Utility Scissors, Physician Grade, Sklar
Merit™ Utility Scissors, Physician Grade, Sklar
Catalog # 10193-690
Supplier:  Sklar
undefined
Merit™ Utility Scissors, Physician Grade, Sklar
Catalog # 10193-690
Supplier:  Sklar
Supplier Number:  97-105
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Length:
    190.5 mm (7½")
  • Material:
    Stainless steel
  • Environmentally Preferable:
  • Cat. no.:
    10193-690
  • Supplier No.:
    97-105
  • Blade Type:
    Angled, Blunt/Blunt
Specifications
About this item
Merit™ Utility scissors are a multipurpose instrument used for cutting non-sterile materials such as dressings, casts, and surgical drapes.
The flattened, wide, blunt tip on the serrated blade greatly reduces the likelihood of skin being nicked if the scissors are used for cutting material that is on the patient, such as clothing or bandages. They should never be used to cut tissue or sutures. These scissors feature fully autoclavable plastic finger ring handles with one ring that is larger than the other.
Merit™ instruments are office-grade products used for routine and in-office minor procedures. These high quality instruments are crafted from Pakistan steel and come with a 10 year guarantee. Merit™ instruments deliver a solution for physicians and practices that require quality, durable instruments.