ContactUsLinkComponent
Boehler Bone Cutting Forceps, OR Grade, Sklar
Boehler Bone Cutting Forceps, OR Grade, Sklar
Catalog # 10195-856
Supplier:  Sklar
undefined
Boehler Bone Cutting Forceps, OR Grade, Sklar
Catalog # 10195-856
Supplier:  Sklar
Supplier Number:  40-4560
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Length:
    146 mm (5¾")
  • Material:
    Stainless steel
  • Tip Configuration:
    Sharp Pointed
  • Curvature:
    Straight
  • Cat. no.:
    10195-856
  • Supplier No.:
    40-4560
Specifications
About this item
Boehler Bone Cutting Forceps are an orthopedic instrument used for cutting bones in a variety of procedures.
The double-action mechanism allows the user to exert less effort while cutting through tougher bone. This product has sharp edges.