To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Length:203 mm (8")
- Material:Stainless Steel
- Cat. no.:10197-470
- Tip Style:Straight, 1 Side Fenestrated
- Supplier No.:66-1240
Specifications
About this item
Spencer Chalazion Forceps are ophthalmologic thumb forceps used to evert the eyelid and expose chalazion for drainage and removal.
This instrument has two circular tips: One is solid and the other is fenestrated. The chalazion on the eyelid is isolated between the two tips and the forceps are tightened by a lock, which pushes the chalazion upwards so the procedure can be completed. The size and consistency of the chalazion are primary factors when choosing which chalazion forceps to use. Larger chalazion, as well as those with hard matter, will require larger incisions. Therefore, forceps with larger tips may be more useful. Similarly, smaller chalazion, or those determined to contain only fluid, may be excised with smaller incisions and the use of forceps with a smaller diameter