Order Entry
Canada
ContactUsLinkComponent
Gill-Hess Iris Forceps, OR Grade, Sklar
Gill-Hess Iris Forceps, OR Grade, Sklar
  10198-088
 :  Sklar
undefined
Gill-Hess Iris Forceps, OR Grade, Sklar
  10198-088
 :  Sklar
 :  66-7240

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Length:
    70 mm (2¾")
  • Material:
    Stainless steel
  • Tip Configuration:
    1x2 Teeth
  • Cat. no.:
    10198-088
  • Tip Style:
    Angled
  • Supplier No.:
    66-7240

 

 

Sklar Gill-Hess Iris Forceps are reusable, Stainless steel and OR grade.

  • Designed for general surgery
  • Ophthalmology
  • Veterinary health

This product is angled, thumb handle style, and 2.75” in length.