Corning® Trace Elements C, Corning
Trace Elements C, 100 ml
Supplier: Corning
: 25-023-CI
Jump to:
Product Details & Documents
Trace Elements C Solution has the same composition as Cleveland's Trace Elements I, and contains the trace elements found in the serum-free medium MCDB301.
Specifications
Specifications
- Description:Trace Elements C
- Volume:100 ml