ContactUsLinkComponent
Corning® Trace Elements B, Corning
Corning® Trace Elements B, Corning
Catalog # CA89422-908
Supplier:  Corning
CAS Number:  
undefined
Corning® Trace Elements B, Corning
Catalog # CA89422-908
Supplier:  Corning
Supplier Number:  25-022-CI
CAS Number:  
Specifications
  • Description:
    Trace Elements B
  • Cat. no.:
    CA89422-908
  • Volume:
    100 ml
  • Supplier No.:
    25-022-CI
Specifications
About this item
Trace Elements B Solution has the same composition as Cleveland's Trace Element I.
The combination of Trace Elements B contains the trace elements of MCDB 104 and of MCDB 301.
This product is proprietary and not available from other major suppliers.