Order Entry
Canada
ContactUsLinkComponent
Human ((13C6)Leu15)-Pth (1-34) Trifluoroacetate
Human ((13C6)Leu15)-Pth (1-34) Trifluoroacetate
Supplier:  Bachem Americas
Certificates
Human ((13C6)Leu15)-Pth (1-34) Trifluoroacetate
Supplier:  Bachem Americas

Specifications

  • Species:
    Human
  • Storage conditions:
    –20 ± 5 °C
  • Protein/peptide name:
    ((¹³C₆)Leu15)-Pth (1-34) Trifluoroacetate
  • Molecular weight:
    4123.71
  • Sequence:
    H-Ser-Val-Ser-Glu-Ile-Gln-Leu-Met-His-Asn-Leu-Gly-Lys-His-[¹³C₆]Leu-Asn-Ser-Met-Glu-Arg-Val-Glu-Trp-Leu-Arg-Lys-Lys-Leu-Gln-Asp-Val-His-Asn-Phe-OH
  • Molecular formula:
    C₁₇₅¹³C₆H₂₉₁N₅₅O₅₁S₂

Specifications

Frequently Bought Together