About this item
TPO is a lineage-specific growth factor produced in the liver, kidney and skeletal muscle. It stimulates the proliferation and maturation of megakaryocytes, and promotes increased circulating levels of platelets in vivo . TPO signals through the c-mpl receptor, and acts as an important regulator of circulating platelets. Human and murine TPO exhibit cross-species reactivity. Recombinant Rat TPO is a fully biologically active 174 amino acid polypeptide (18.7 kDa), which contains the erythropoietin-like domain of the full length TPO protein.
- Animal-free
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokines and Growth Factors
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Rat
- Storage conditions:Store at –20 °C prior to reconstitution.
- Endotoxin content:>0.1 ng/μg
- Biological activity:Biological Activity tests can vary. Please refer to the Product CoA for information.
- Form:Lyophilized
- Gene ID:81811 P49745
- Reconstitution instructions:Reconstitution instructions can vary. Please refer to the product CoA for info.
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:MGDF|c-MPL Ligand|Megakaryocyte colony-stimulating factor|Thrombopoietin
- Protein/peptide name:TPO
- Purity:0.98
- Molecular weight:18.7 kDa
- Sequence:SPVPPACDPR LLNKLLRDSY LLHSRLSQCP DVNPLSIPVL LPAVDFSLGE WKTQTEQSKA QDILGAVSLL LEGVMAARGQ LEPSCLSSLL GQLSGQVRLL LGALQGLLGT QLPPQGRTTA HKDPSALFLS LQQLLRGKVR FLLLVEGPAL CVRRTLPTTA VPSRTSQLLT LNKF
- Endotoxin level:Low
- Formulation:Formulation can vary. Please refer to product CoA for information.
- Shipping temperature:Ambient