Order Entry
Canada
Orders LinkContactUsLinkComponent
<i>Staphylococcus</i> Recombinant Glu-C (from <i>E. coli</i>)

Staphylococcus Recombinant Glu-C (from E. coli)

Supplier:   Peprotech
<i>Staphylococcus</i> Recombinant Glu-C (from <i>E. coli</i>)
Supplier:  Peprotech
Certificates
Staphylococcus Recombinant Glu-C (from E. coli)

4 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Proteases (also called Proteolytic Enzymes, Peptidases, or Proteinases) are enzymes that hydrolyze the amide bonds within proteins or peptides. Most proteases act in a specific manner, hydrolyzing bonds at, or adjacent to, specific residues, or a specific sequence of residues contained within the substrate protein or peptide. Proteases play an important role in most diseases and biological processes, including prenatal and postnatal development, reproduction, signal transduction, immune response, various autoimmune and degenerative diseases, and cancer. They are also an important research tool, as they are frequently used in the analysis and production of proteins. Glu-C cleaves at the Carboxyl side of E (can also cleave D under certain conditions). Recombinant Staphylococcus Glu-C is a 28.8 kDa protease consisting of 266 amino acid residues.

Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 10781-372
  • 10781-374
  • 10781-376
  • 10781-378
  • Protein/peptide name:
  • Glu-C
  • Glu-C
  • Glu-C
  • Glu-C
  • Protein synonyms:
  • V8 Protease
  • V8 Protease
  • V8 Protease
  • V8 Protease
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Protein Function:
  • Cytokines and Growth Factors
  • Cytokines and Growth Factors
  • Cytokines and Growth Factors
  • Cytokines and Growth Factors
  • Species:
  • Staphylococcus
  • Staphylococcus
  • Staphylococcus
  • Staphylococcus
  • Source:
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Gene ID:
  • 3920389 Q2FZL2
  • 3920389 Q2FZL2
  • 3920389 Q2FZL2
  • 3920389 Q2FZL2
  • Sequence:
  • LPNNDRHQIT DTTNGHYAPV TYIQVEAPTG TFIASGVVVG KDTLLTNKHV VDATHGDPHA LKAFPSAINQ DNYPNGGFTA EQITKYSGEG DLAIVKFSPN EQNKHIGEVV KPATMSNNAE TQVNQNITVT GYPGDKPVAT MWESKGKITY LKGEAMQYDL STTGGNSGSP VFNEKNEVIG IHWGGVPNEF NGAVFINENV RNFLKQNIED IHFANDDQPN NPDNPDNPNN PDNPNNPDEP NNPDNPNNPD NPDNGDNNNS DNPDAA
  • LPNNDRHQIT DTTNGHYAPV TYIQVEAPTG TFIASGVVVG KDTLLTNKHV VDATHGDPHA LKAFPSAINQ DNYPNGGFTA EQITKYSGEG DLAIVKFSPN EQNKHIGEVV KPATMSNNAE TQVNQNITVT GYPGDKPVAT MWESKGKITY LKGEAMQYDL STTGGNSGSP VFNEKNEVIG IHWGGVPNEF NGAVFINENV RNFLKQNIED IHFANDDQPN NPDNPDNPNN PDNPNNPDEP NNPDNPNNPD NPDNGDNNNS DNPDAA
  • LPNNDRHQIT DTTNGHYAPV TYIQVEAPTG TFIASGVVVG KDTLLTNKHV VDATHGDPHA LKAFPSAINQ DNYPNGGFTA EQITKYSGEG DLAIVKFSPN EQNKHIGEVV KPATMSNNAE TQVNQNITVT GYPGDKPVAT MWESKGKITY LKGEAMQYDL STTGGNSGSP VFNEKNEVIG IHWGGVPNEF NGAVFINENV RNFLKQNIED IHFANDDQPN NPDNPDNPNN PDNPNNPDEP NNPDNPNNPD NPDNGDNNNS DNPDAA
  • LPNNDRHQIT DTTNGHYAPV TYIQVEAPTG TFIASGVVVG KDTLLTNKHV VDATHGDPHA LKAFPSAINQ DNYPNGGFTA EQITKYSGEG DLAIVKFSPN EQNKHIGEVV KPATMSNNAE TQVNQNITVT GYPGDKPVAT MWESKGKITY LKGEAMQYDL STTGGNSGSP VFNEKNEVIG IHWGGVPNEF NGAVFINENV RNFLKQNIED IHFANDDQPN NPDNPDNPNN PDNPNNPDEP NNPDNPNNPD NPDNGDNNNS DNPDAA
  • Endotoxin level:
  • Low
  • Low
  • Low
  • Low
  • Endotoxin content:
  • >0.1 ng/μg
  • >0.1 ng/μg
  • >0.1 ng/μg
  • >0.1 ng/μg
  • Biological activity:
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Purity:
  • 0.95
  • 0.95
  • 0.95
  • 0.95
  • Form:
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Formulation:
  • Formulation can vary. Please refer to product CoA for information.
  • Formulation can vary. Please refer to product CoA for information.
  • Formulation can vary. Please refer to product CoA for information.
  • Formulation can vary. Please refer to product CoA for information.
  • Molecular weight:
  • 28.8 kDa
  • 28.8 kDa
  • 28.8 kDa
  • 28.8 kDa
  • Reconstitution instructions:
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Storage conditions:
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Shipping temperature:
  • Ambient
  • Ambient
  • Ambient
  • Ambient

Specifications

Product Family Options

Product Information

  • Size
    Origin
    Availability
    Price
  • 50 μg
    US
    Log in for availability
    Log in for availability
    Each (50µG)
    Each (50µG) - $248.04
    $248.04
    Each (50µG)
       $248.04
    Minimum order quantity is 1

    Supplier Number: 450-46-50UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10781-372
    Protein/peptide name
    Glu-C
    Protein synonyms
    V8 Protease
    Protein/peptide type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Staphylococcus
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    3920389 Q2FZL2
    Sequence
    LPNNDRHQIT DTTNGHYAPV TYIQVEAPTG TFIASGVVVG KDTLLTNKHV VDATHGDPHA LKAFPSAINQ DNYPNGGFTA EQITKYSGEG DLAIVKFSPN EQNKHIGEVV KPATMSNNAE TQVNQNITVT GYPGDKPVAT MWESKGKITY LKGEAMQYDL STTGGNSGSP VFNEKNEVIG IHWGGVPNEF NGAVFINENV RNFLKQNIED IHFANDDQPN NPDNPDNPNN PDNPNNPDEP NNPDNPNNPD NPDNGDNNNS DNPDAA
    Endotoxin level
    Low
    Endotoxin content
    >0.1 ng/μg
    Biological activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.95
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular weight
    28.8 kDa
    Reconstitution instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage conditions
    Store at –20 °C prior to reconstitution.
    Shipping temperature
    Ambient
  • 250 μg
    US
    Log in for availability
    Log in for availability
    Each (250µG)
    Each (250µG) - $541.77
    $541.77
    Each (250µG)
       $541.77
    Minimum order quantity is 1

    Supplier Number: 450-46-250UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10781-374
    Protein/peptide name
    Glu-C
    Protein synonyms
    V8 Protease
    Protein/peptide type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Staphylococcus
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    3920389 Q2FZL2
    Sequence
    LPNNDRHQIT DTTNGHYAPV TYIQVEAPTG TFIASGVVVG KDTLLTNKHV VDATHGDPHA LKAFPSAINQ DNYPNGGFTA EQITKYSGEG DLAIVKFSPN EQNKHIGEVV KPATMSNNAE TQVNQNITVT GYPGDKPVAT MWESKGKITY LKGEAMQYDL STTGGNSGSP VFNEKNEVIG IHWGGVPNEF NGAVFINENV RNFLKQNIED IHFANDDQPN NPDNPDNPNN PDNPNNPDEP NNPDNPNNPD NPDNGDNNNS DNPDAA
    Endotoxin level
    Low
    Endotoxin content
    >0.1 ng/μg
    Biological activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.95
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular weight
    28.8 kDa
    Reconstitution instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage conditions
    Store at –20 °C prior to reconstitution.
    Shipping temperature
    Ambient
  • 500 μg
    US
    Log in for availability
    Log in for availability
    Each (500µG)
    Each (500µG) - $1,000.01
    $1,000.01
    Each (500µG)
       $1,000.01
    Minimum order quantity is 1

    Supplier Number: 450-46-500UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10781-376
    Protein/peptide name
    Glu-C
    Protein synonyms
    V8 Protease
    Protein/peptide type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Staphylococcus
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    3920389 Q2FZL2
    Sequence
    LPNNDRHQIT DTTNGHYAPV TYIQVEAPTG TFIASGVVVG KDTLLTNKHV VDATHGDPHA LKAFPSAINQ DNYPNGGFTA EQITKYSGEG DLAIVKFSPN EQNKHIGEVV KPATMSNNAE TQVNQNITVT GYPGDKPVAT MWESKGKITY LKGEAMQYDL STTGGNSGSP VFNEKNEVIG IHWGGVPNEF NGAVFINENV RNFLKQNIED IHFANDDQPN NPDNPDNPNN PDNPNNPDEP NNPDNPNNPD NPDNGDNNNS DNPDAA
    Endotoxin level
    Low
    Endotoxin content
    >0.1 ng/μg
    Biological activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.95
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular weight
    28.8 kDa
    Reconstitution instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage conditions
    Store at –20 °C prior to reconstitution.
    Shipping temperature
    Ambient
  • 1 mg
    US
    Log in for availability
    Log in for availability
    Each (1mg)
    Each (1mg) - $1,665.19
    $1,665.19
    Each (1mg)
       $1,665.19
    Minimum order quantity is 1

    Supplier Number: 450-46-1MG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10781-378
    Protein/peptide name
    Glu-C
    Protein synonyms
    V8 Protease
    Protein/peptide type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Staphylococcus
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    3920389 Q2FZL2
    Sequence
    LPNNDRHQIT DTTNGHYAPV TYIQVEAPTG TFIASGVVVG KDTLLTNKHV VDATHGDPHA LKAFPSAINQ DNYPNGGFTA EQITKYSGEG DLAIVKFSPN EQNKHIGEVV KPATMSNNAE TQVNQNITVT GYPGDKPVAT MWESKGKITY LKGEAMQYDL STTGGNSGSP VFNEKNEVIG IHWGGVPNEF NGAVFINENV RNFLKQNIED IHFANDDQPN NPDNPDNPNN PDNPNNPDEP NNPDNPNNPD NPDNGDNNNS DNPDAA
    Endotoxin level
    Low
    Endotoxin content
    >0.1 ng/μg
    Biological activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.95
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular weight
    28.8 kDa
    Reconstitution instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage conditions
    Store at –20 °C prior to reconstitution.
    Shipping temperature
    Ambient