Order Entry
Canada
ContactUsLinkComponent
Human Recombinant GDNF (from E. coli)
Human Recombinant GDNF (from E. coli)
Supplier:  Peprotech
Certificates
Human Recombinant GDNF (from E. coli)
Supplier:  Peprotech

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C prior to reconstitution.
  • Endotoxin content:
    >0.1 ng/μg
  • Biological activity:
    Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Form:
    Lyophilized
  • Gene ID:
    2668 P39905
  • Reconstitution instructions:
    Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Protease-free:
    Y
  • Protein synonyms:
    Glial-Derived Neurotrophic Factor|ATF (Astrocyte-derived Trophic Factor)
  • Protein/peptide name:
    GDNF
  • Purity:
    0.98
  • Molecular weight:
    30.4 kDa
  • Sequence:
    MSPDKQMAVL PRRERNRQAA AANPENSRGK GRRGQRGKNR GCVLTAIHLN VTDLGLGYET KEELIFRYCS GSCDAAETTY DKILKNLSRN RRLVSDKVGQ ACCRPIAFDD DLSFLDDNLV YHILRKHSAK RCGCI
  • Endotoxin level:
    Low
  • Formulation:
    Formulation can vary. Please refer to product CoA for information.
  • Shipping temperature:
    Ambient

Specifications

Related Information

Frequently Bought Together